T4 DNA Polymerase, New England Biolabs
Supplier: New England Biolabs (NEB)
M0203L M0203S
CA101228-184EA 419 CAD
CA101228-184 CA101228-186
T4 DNA Polymerase, New England Biolabs
Nucleic Acid Reagents Nucleic Acid Modification and Labelling
T4 DNA Polymerase catalyzes the synthesis of DNA in the 5´→ 3´ direction and requires the presence of template and primer
This enzyme has a 3´→ 5´ exonuclease activity which is much more active than that found in DNA Polymerase I (E. coli). Unlike E. coli DNA Polymerase I, T4 DNA Polymerase does not have a 5´→ 3´ exonuclease function.