T4 DNA Ligase, New England Biolabs
Supplier: New England Biolabs (NEB)
M0202L
M0202M
M0202T
M0202S
CA101228-176EA
410
CAD
CA101228-176
CA101228-178
CA101228-182
CA101228-180
T4 DNA Ligase, New England Biolabs
Nucleic Acid Reagents
Cloning Kits and Ligation
Catalyzes the formation of a phosphodiester bond between juxtaposed 5' phosphate and 3' hydroxyl termini in duplex DNA or RNA.
This enzyme will join blunt end and cohesive end termini as well as repair single stranded nicks in duplex DNA and some DNA/RNA hybrids. T4 DNA ligase will seal nicks for these DNA substrates.