ShortCut RNase III, New England Biolabs
Supplier: New England Biolabs (NEB)
M0245L
M0245S
CA101227-632EA
888
CAD
CA101227-632
CA101227-634
ShortCut RNase III, New England Biolabs
Enzymes
ShortCut® RNase III, used with its manganese-containing reaction buffer, converts long double-stranded RNA into a heterogeneous mix of short (18–25 bp) interfering RNAs (siRNA) suitable for RNA interference in mammalian cells
1.5 units (1 µl) of ShortCut RNase III is sufficient to convert 1 µg of dsRNA into siRNA suitable for RNA interference in mammalian cells.