Human RNase Inhibitor (from Placenta)
Supplier: New England Biolabs (NEB)
M0307L M0307S
CA101227-650EA 569 CAD
CA101227-650 CA101227-652
Human RNase Inhibitor (from Placenta)
Proteins and Peptides
RNase Inhibitor, Human Placenta is a recombinant human placental protein which specifically inhibits ribonucleases (RNases) A, B and C
It is not effective against RNase 1, RNase T1, S1 Nuclease, RNase H or RNase from Aspergillus. In addition, no inhibition of polymerase activity is observed when RNase Inhibitor, Human Placenta is used with Taq DNA Polymerase, AMV or M-MuLV Reverse Transcriptases, or Phage RNA Polymerases (SP6, T7, or T3). The 50 kDa protein inhibits RNases by binding noncovalently in a 1:1 ratio with an association constant greater than 10^14.