RNase HII
Supplier: New England Biolabs (NEB)
M0288L M0288S
CA101444-756EA 521 CAD
CA101444-756 CA101444-758
RNase HII
Enzymes
Ribonuclease HII (RNase HII) is an endoribonuclease that preferentially nicks 5´ to a ribonucleotide within the context of a DNA duplex
The enzyme leaves 5´ phosphate and 3´ hydroxyl ends. RNase HII will also nick at multiple sites along the RNA portion of an Okazaki fragment.