RNase H
Supplier: New England Biolabs (NEB)
M0297L M0297S
CA101228-364EA 499 CAD
CA101228-364 CA101228-366
RNase H
Enzymes
Ribonuclease H (RNase H) is an endoribonuclease which specifically hydrolyzes the phosphodiester bonds of RNA which is hybridized to DNA
This enzyme does not digest single- or double-stranded DNA.