Ribonucleotide Solution Set, New England Biolabs
Supplier: New England Biolabs (NEB)
N0450L
N0450S
CA101641-038EA
499
CAD
CA101641-038
CA101641-040
Ribonucleotide Solution Set, New England Biolabs
Nucleic Acid Reagents
Nucleotides
The Ribonucleotide Solution Set (NTP Set) consists of four separate solutions of ATP, GTP, CTP and UTP, pH 7.5, as sodium salts
Each nucleotide is supplied at 100 mM. The NTPs are also available as a Ribonucleotide Solution Mix.