Ribonucleotide Solution Mix, New England Biolabs
Supplier: New England Biolabs (NEB)
N0466L N0466S
CA101228-422EA 505 CAD
CA101228-422 CA101228-424
Ribonucleotide Solution Mix, New England Biolabs
Nucleic Acid Reagents Nucleotides
Ribonucleotide Solution Mix is a buffered equimolar solution of ribonucleotide triphosphates: rATP, rCTP, rGTP and rUTP
Each nucleotide is supplied at a concentration of 25 mM (total rNTP concentration equals 100 mM). The NTPs are also available as Ribonucleotide Solution Set.
Order Now
SPECIFICATIONS
Environmentally Preferable
Application
Learn more