RecA
Supplier: New England Biolabs (NEB)
M0249S M0249L
CA101228-272EA 123 CAD
CA101228-272 CA101641-566
RecA
Proteins and Peptides
E. coli RecA is necessary for genetic recombination, reactions involving DNA repair and UV-induced mutagenesis. RecA promotes the autodigestion of the lexA repressor, umuD protein and lambda repressor. Cleavage of LexA derepresses more than 20 genes. In vitro studies indicate that in the presence of ATP, RecA promotes the strand exchange of single-strand DNA fragments with homologous duplex DNA. The reaction has three distinct steps: (i) RecA polymerizes on the single-strand DNA, (ii) the nucleoprotein filament binds the duplex DNA and searches for a homologous region, (iii) the strands are exchanged.