Random Primer Mix, New England Biolabs
Supplier: New England Biolabs
S1330S S1330S
CA101641-208EA 165 CAD
CA101641-208 101641-208
Random Primer Mix, New England Biolabs
Nucleic Acid Reagents Primers and Probes
Random priming is widely used in the first strand cDNA synthesis
Random Primer Mix is a ready-to-use optimized mixture of hexamers and anchored-dT primer (dT23VN). A mixture of hexamers and anchored-dT primer provides even and consistent coverage of the RNA template population across a wide range of RNA template concentration. In contrast, traditional random priming by hexamer has poor coverage of 3´ end of RNA templates.