QUICK-LOAD® DNA Ladder, 100 bp, New England Biolabs
Supplier: New England Biolabs (NEB)
N0467S N0467L
CA101227-136EA 149 CAD
CA101227-136 CA101449-370
QUICK-LOAD® DNA Ladder, 100 bp, New England Biolabs
Electrophoresis Reagents Molecular Weight Markers
Quick-Load® 100 bp DNA Ladder is a pre-mixed, ready-to-load molecular weight marker containing bromophenol blue as a tracking dye
The DNA Ladder consists of proprietary plasmids which are digested to completion with appropriate restriction enzymes to yield 12 bands suitable for use as molecular weight standards for agarose gel electrophoresis. The digested DNA includes fragments ranging from 100-1,517 base pairs. The 500 and 1,000 base pair bands have increased intensity to serve as reference bands.