QUICK-LOAD® DNA Ladder, 1 kb, New England Biolabs
Supplier: New England Biolabs (NEB)
N0468S N0468L
CA101228-426EA 112 CAD
CA101228-426 CA101449-372
QUICK-LOAD® DNA Ladder, 1 kb, New England Biolabs
Electrophoresis Reagents Molecular Weight Markers
Quick-Load® 1 kb DNA Ladder is a premixed, ready-to-load molecular weight marker containing bromophenol blue as a tracking dye
The DNA Ladder consists of proprietary plasmids which are digested to completion with appropriate restriction enzymes to yield 10 bands suitable for use as molecular weight standards for agarose gel electrophoresis. The digested DNA includes fragments ranging from 0.5-10.0 kilobases. The 3.0 kb fragment has increased intensity to serve as reference bands.