Yeast Pyrophosphatase
Supplier: New England Biolabs (NEB)
M2403S M2403L M2403S
101417-998EA 126.4 CAD
101417-998 CA101417-996 CA101417-998
Yeast Pyrophosphatase
Enzymes
Inorganic pyrophosphatase (PPase) catalyzes the hydrolysis of inorganic pyrophosphate to form orthophosphate
Pyrophosphatase, Inorganic (yeast) is prepared from an E. coli strain containing a genetic fusion of the Saccharomyces cerevisiae ppa gene and the gene coding for Mycobacterium xenopi GyrA intein. The final product has been cleaved and purified from the intein fusion partner. Developed by the Biohelix Corporation, and produced at New England Biolabs.