PvuII

Supplier: New England Biolabs (NEB)

R0151M R0151T
CA101228-824EA 426 CAD
CA101228-824 CA101228-828
PvuII
Nucleic Acid Reagents Restriction Enzymes

An E.coli strain that carries the PvuII gene from Proteus vulgaris (ATCC 13315).

Order Now

SPECIFICATIONS

Environmentally Preferable
Application
Enzyme name PvuII
Source

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR