PUREXPRESS® In Vitro Protein Synthesis Kit, New England Biolabs
Supplier: New England Biolabs (NEB)
E6800L E6800S
CA102715-964EA 3932 CAD
CA102715-964 CA101446-494
PUREXPRESS® In Vitro Protein Synthesis Kit, New England Biolabs
Protein Expression Protein Expression Systems
A rapid method for gene expression analysis, PURExpress® is a novel cell-free transcription/translation system reconstituted from the purified components necessary for E.coli translation.
The relative nuclease-free and protease-free nature of the PURExpress platform preserves the integrity of DNA and RNA templates/ complexes and results in proteins that are free of modification and degradation. Transcription and translation are carried out in a one-step reaction, and require the mixing of only two tubes. With results available in a few hours, PURExpress saves valuable laboratory time and is ideal for high throughput technologies.