pUC19 Vector, New England Biolabs
Supplier: New England Biolabs (NEB)
N3041L N3041S
CA101228-478EA 516 CAD
CA101228-478 CA101228-480
pUC19 Vector, New England Biolabs
Vectors, Plasmids and Libraries
pUC19 is a commonly used plasmid cloning vector in E.coli
The molecule is a small double-stranded circle, 2686 base pairs in length, and has a high copy number. pUC19 carries a 54 base-pair multiple cloning site polylinker that contains unique sites for 13 different hexanucleotide-specific restriction endonucleases.
Order Now
SPECIFICATIONS
Environmentally Preferable