ProtoScript M-MuLV First Strand cDNA Synthesis Kit, New England Biolabs
Supplier: New England Biolabs (NEB)
E6300L E6300S
CA101640-908EA 1096 CAD
CA101640-908 CA101640-910
ProtoScript M-MuLV First Strand cDNA Synthesis Kit, New England Biolabs
Nucleic Acid Reagents RT-PCR and cDNA Reagents
ProtoScript® First Strand cDNA Synthesis Kit features two optimized mixes, M-MuLV Enzyme Mix and M-MuLV Reaction Mix
The kit also contains two optimized primers for reverse transcription and nuclease-free water. An anchored oligo-dT primer [d(T)23VN] forces the primer to anneal to the beginning of the polyA tail. The optimized Random Primer Mix provides random and consistent priming sites covering the entire RNA template including both mRNAs and non-polyadenylated RNAs. The first strand cDNA product generated is more than 10 kb.
M-MuLV Enzyme Mix combines M-MuLV Reverse Transcriptase and Murine RNase Inhibitor, while M-MuLV Reaction Mix contains dNTPs and an optimized buffer.