Native PNGase F
Supplier: New England Biolabs (NEB)
P0704L P0704S
CA101227-724EA 1361 CAD
CA101227-724 CA101227-726
Native PNGase F
Enzymes
Peptide-N-Glycosidase F, also known as PNGase F, is an amidase which cleaves between the innermost GlcNAc and asparagine residues of high mannose, hybrid and complex oligosaccharides from N-linked glycoproteins
A glycerol-free version of PNGaseF is also offered for HPLC methods.