Phusion Hot Start Flex DNA Polymerase, New England Biolabs
Supplier: New England Biolabs (NEB)
M0535L
M0535S
CA102500-128EA
911
CAD
CA102500-128
CA102500-130
Phusion Hot Start Flex DNA Polymerase, New England Biolabs
Nucleic Acid Reagents
End-point PCR Enzymes and Kits
Phusion Hot Start Flex DNA Polymerase offers robust, high fidelity performance and room temperature reaction setup
The addition of an aptamer-based inhibitor allows room temperature reaction setup. Phusion Hot Start Flex DNA Polymerase is supplied with standard 5X Phusion HF Buffer, as well as 5X Phusion GC Buffer, which can be used for complex or GC-rich templates.