Phusion Hot Start Flex 2X Master Mix, New England Biolabs
Supplier: New England Biolabs (NEB)
M0536L M0536S
CA102500-124EA 1290 CAD
CA102500-124 CA102500-126
Phusion Hot Start Flex 2X Master Mix, New England Biolabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Phusion Hot Start Flex 2X Master Mix offers robust high fidelity performance and room temperature reaction setup in a convenient master mix format
The addition of an aptamer-based inhibitor allows room temperature reaction setup. The convenient master mix formulation is supplied at a 2X concentration and contains dNTPs and Mg++, requiring only the addition of primers and DNA template for robust amplification. Reactions can also be optimized using the provided DMSO solution.