Phusion High-Fidelity PCR Master Mix with HF® Buffer, New England Biolabs
Supplier: New England Biolabs (NEB)
M0531L
M0531S
CA101641-016EA
1185
CAD
CA101641-016
CA101641-018
Phusion High-Fidelity PCR Master Mix with HF® Buffer, New England Biolabs
Nucleic Acid Reagents
End-point PCR Enzymes and Kits
Phusion High-Fidelity PCR Master Mix with HF Buffer is a 2X master mix consisting of Phusion DNA Polymerase, deoxynucleotides and reaction buffer that has been optimized and includes MgCl2
All that is required is the addition of template, primers and water.