Phusion High-Fidelity PCR Master Mix with GC Buffer, New England Biolabs
Supplier: New England Biolabs (NEB)
M0532L M0532S
CA101641-020EA 1185 CAD
CA101641-020 CA101641-022
Phusion High-Fidelity PCR Master Mix with GC Buffer, New England Biolabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Phusion High-Fidelity PCR Master Mix with GC Buffer offers robust performance and can be used for all PCR applications
It is a 2X master mix consisting of Phusion DNA Polymerase, deoxynucleotides and reaction buffer that has been optimized and includes MgCl2. All that is required is the addition of template, primers and water.