PhiX174 DNA-Hae III Digest, New England Biolabs
Supplier: New England Biolabs (NEB)
N3026L
N3026S
CA101228-462EA
549
CAD
CA101228-462
CA101228-464
PhiX174 DNA-Hae III Digest, New England Biolabs
Electrophoresis Reagents
Molecular Weight Markers
The HaeIII digest of ΦX174 yields 11 fragments suitable for use as molecular weight standards for agarose gel electrophoresis
Comes supplied with 1 vial of Gel Loading Dye, Purple (6X), no SDS.