Phi29 DNA Polymerase, New England Biolabs
Supplier: New England Biolabs (NEB)
M0269L
M0269S
CA101228-322EA
366
CAD
CA101228-322
CA101228-324
Phi29 DNA Polymerase, New England Biolabs
Nucleic Acid Reagents
End-point PCR Enzymes and Kits
phi29 DNA Polymerase is the replicative polymerase from the Bacillus subtilis phage phi29 (Φ29)
This polymerase has exceptional strand displacement and processive synthesis properties. The polymerase has an inherent 3´→5' proofreading exonuclease activity.