ONETAQ® QUICK-LOAD® 2X Master Mix with Standard Buffer, New England Biolabs
Supplier: New England Biolabs (NEB)
M0486L M0486S
CA101640-996EA 333 CAD
CA101640-996 CA101640-998
ONETAQ® QUICK-LOAD® 2X Master Mix with Standard Buffer, New England Biolabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
OneTaq® Quick-Load® 2X Master Mix with Standard Buffer is an optimized, ready-to-use blend of Taq and Deep VentR ™ DNA Polymerases ideally suited to routine PCR applications from a variety of templates, including pure DNA solutions, bacterial colonies and cDNA products.
The 3´→5´ exonuclease activity of Deep Vent DNA Polymerase increases the fidelity and robust amplification of Taq DNA Polymerase. The convenient quick-load master mix formulation contains dNTPs, MgCl2, buffer components and stabilizers as well as two commonly used tracking dyes for DNA gels. On a 1% agarose gel in 1X TBE, Xylene Cyanol FF migrates at ~4 kb and Tartrazine migrates at ~10 bp. Both dyes are present in concentrations that do not mask comigrating DNA bands.
Order Now
SPECIFICATIONS
Environmentally Preferable
Application
Learn more