ONETAQ® Hot Start QUICK-LOAD® 2X Master Mix with GC Buffer, New England Biolabs
Supplier: New England Biolabs (NEB)
OneTaq® Hot Start Quick-Load® 2X Master Mix with GC Buffer is an optimized, ready-to-use blend of Taq and Deep VentR™ DNA Polymerases combined with an aptamer-based inhibitor.
This enzyme blend is ideally suited to PCR applications from GC-rich templates, including pure DNA solutions, bacterial colonies, and cDNA products. The 3´→5´ exonuclease activity of Deep Vent DNA Polymerase increases the fidelity and robust amplification of Taq DNA Polymerase. The hot start nature of the enzyme offers convenience with decreased interference from primer dimers and secondary products. The convenient master mix formulation contains dNTPs, MgSO4, buffer components and stabilizers as well as two commonly used tracking dyes for DNA gels. On a 1% agarose gel in 1X TBE, Xylene Cyanol FF migrates at ~4 kb and Tartrazine migrates at ~10 bp. Both dyes are present in concentrations that do not mask co-migrating DNA bands.