ONETAQ® Hot Start 2X Master Mix with GC Buffer, New England Biolabs
Supplier: New England Biolabs (NEB)
M0485L M0485S
CA101640-992EA 552 CAD
CA101640-992 CA101640-994
ONETAQ® Hot Start 2X Master Mix with GC Buffer, New England Biolabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
OneTaq® Hot Start 2X Master Mix with GC Buffer is an optimized blend of Taq and Deep VentR ™ DNA Polymerases combined with an aptamer-based inhibitor.
This enzyme blend is ideally suited to PCR applications from GC-rich templates, including pure DNA solutions, bacterial colonies and cDNA products. The 3´→5´ exonuclease activity of Deep Vent DNA Polymerase increases the fidelity and robust amplification of Taq DNA Polymerase. The hot start nature of the enzyme offers convenience with decreased interference from primer dimers and secondary products. The convenient master mix formulation contains dNTPs, MgSO4, buffer components and stabilizers, requiring only the addition of primers and DNA template for robust amplification.