ONETAQ® DNA Polymerase, New England Biolabs
Supplier: New England Biolabs (NEB)
OneTaq DNA Polymerase is an optimized blend of Taq and Deep VentR™ DNA polymerases for use with routine and difficult PCR experiments
The 3´→ 5´ exonuclease activity of Deep Vent DNA Polymerase increases the fidelity and robust amplification of Taq DNA Polymerase. The OneTaq Reaction Buffers and High GC Enhancer have been formulated for robust yields with minimal optimization, regardless of a template’s GC content. OneTaq is provided with Standard Reaction Buffer, GC Reaction Buffer and High GC Enhancer.