ONETAQ® 2X Master Mix with Standard Buffer, New England Biolabs
Supplier: New England Biolabs (NEB)
M0482L M0482S
CA101640-980EA 321 CAD
CA101640-980 CA101640-982
ONETAQ® 2X Master Mix with Standard Buffer, New England Biolabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
OneTaq® 2X Master Mix with Standard Buffer is an optimized blend of Taq and Deep VentR™ DNA Polymerases ideally suited to routine PCR applications from a variety of templates including pure DNA solutions, bacterial colonies, and cDNA products
The 3´→5´ exonuclease activity of Deep Vent DNA Polymerase increases the fidelity and robust amplification of Taq DNA Polymerase. The convenient master mix formulation contains dNTPs, MgCl2, buffer components and stabilizers, requiring only the addition of primers and DNA template for robust amplification.