Alpha 2-3,6,8 Neuraminidase
Supplier: New England Biolabs (NEB)
P0720L
P0720S
CA101228-544EA
488
CAD
CA101228-544
CA101228-546
Alpha 2-3,6,8 Neuraminidase
Enzymes
Neuraminidase is the common name for Acetyl-neuraminyl hydrolase (Sialidase)
This Neuraminidase catalyzes the hydrolysis of α2-3, α2-6, and α2-8 linked N-acetyl-neuraminic acid residues from glycoproteins and oligosaccharides.