NEBNEXT® mRNA Second Strand Synthesis Module, New England Biolabs
Supplier: New England Biolabs (NEB)
B6117S
E6111L
E6111S
CA101640-776EA
54
CAD
CA101640-776
CA101640-872
CA101640-874
NEBNEXT® mRNA Second Strand Synthesis Module, New England Biolabs
Nucleic Acid Reagents
Next Generation Sequencing Reagents
The NEBNext Second Strand Synthesis Module has been optimized to generate double stranded cDNA from first strand cDNA
- NEBNext Second Strand Synthesis (dNTP-free) reaction buffer
The NEBNext Second Strand Synthesis Module is provided as a master mix to maximize efficiency and convenience in RNA sample preparation workflows.