NEBNEXT® End Repair Module, New England Biolabs
Supplier: New England Biolabs (NEB)
E6050L
E6050S
CA101640-832EA
641
CAD
CA101640-832
CA101640-834
NEBNEXT® End Repair Module, New England Biolabs
Nucleic Acid Reagents
Next Generation Sequencing Reagents
The NEBNext® End Repair Module has been optimized to repair DNA fragmented by nebulization, acoustic shearing, or nucleases
Fragmented DNA is converted by the NEBNext End Repair Module to blunt ended DNA having 5´-phosphates and 3´-hydroxyl. The NEBNext End Repair Module is provided as a master mix to maximize efficiency and convenience in DNA sample preparation workflows.