NEBNext® NGS DNA Library Preparation for Illumina, Kits and Products, New England Biolabs
Supplier: New England Biolabs (NEB)
E7600S B0349S M6630L M6630S E7630S E7630L M0544L M0544S B6118S E2612L E7645L E7645S E7335L E7335S E7500L E7500S E2612S E7600S B0349S M6630L M6630S E7630S E7630L M0544L M0544S B6118S E2612L E7645L E7645S M0348L M0348S
102877-580EA 807.38 CAD
102877-580 102877-608 102902-480 102902-482 102942-190 102942-192 102969-088 102969-090 102969-094 102969-098 102969-108 102969-110 CA102500-098 CA102500-100 CA102715-932 CA102715-934 CA102855-164 CA102877-572 CA102877-574 CA102877-580 CA102877-608 CA102902-480 CA102902-482 CA102942-190 CA102942-192 CA102969-088 CA102969-090 CA102969-094 CA102969-098 CA102969-108 CA102969-110 CA101640-952 CA101640-954
NEBNext® NGS DNA Library Preparation for Illumina, Kits and Products, New England Biolabs
Nucleic Acid Reagents Next Generation Sequencing Reagents
The NEBNext Ultra II Q5® Master Mix is a new formulation of Q5 DNA polymerase that has been optimized for robust, high-fidelity amplification of next-generation sequencing (NGS) libraries.
  • Fast, streamlined workflow and novel master mixes
  • High yield preparation of high quality libraries
  • Functionally validated by preparation of a library
This new formulation further improves the uniformity of amplification of libraries, including superior performance with GC-rich regions
12-fold higher than that of Pfu DNA Polymerase), resulting in ultra-low error rates. The NEBNext Ultra II Q5 Master Mix is an aptamer-based hot start formulation that allows convenient room temperature reaction set up. The convenient 2X master mix format contains dNTPs, Mg++ and a proprietary buffer. NEBNext Ultra II Q5 Master Mix is also included in the NEBNext Ultra II DNA Library Prep Kit for Illumina®.
Get more of what you need, with the highest library yields. Use these products to generate high quality libraries even when you have only limited amounts of DNA, with inputs as low as 500 pg (Ultra II). Prepare libraries from ALL of your samples, including GC-rich targets and FFPE DNA samples. Improve yields and quality for target enrichment applications. Save time with streamlined workflows, reduced hands-on time, and automation compatibility.
Q5 High-Fidelity DNA Polymerase is fused to a processivity-enhancing Sso7d domain providing the highest fidelity available (
Certifications: In addition to the extensive QCs on individual kit components, all NEBNext kits for Illumina are functionally validated by preparation of a library, followed by Illumina sequencing.
Ordering information: Original NEBNext Ultra kits and sets of individual reagents are also available.