NEB®Express Competent E.coli (High Efficiency), New England Biolabs
Supplier: New England Biolabs (NEB)
C2523H C2523I
CA101417-888EA 358 CAD
CA101417-888 CA101417-890
NEB®Express Competent E.coli (High Efficiency), New England Biolabs
Competent Cells
A versatile non-T7 expression E. coli strain
NEB Express is the recommended host strain for pMAL protein fusion and purification system.