NEB®5-alpha Competent E.coli (High Efficiency), New England Biolabs
Supplier: New England Biolabs (NEB)
C2987H C2987I C2987P C2987R C2987U
CA101417-898EA 314 CAD
CA101417-898 CA101417-900 CA102855-190 CA103218-956 CA103218-958
NEB®5-alpha Competent E.coli (High Efficiency), New England Biolabs
Competent Cells
NEB 5-alpha Competent E. coli (High Efficiency) is a derivative of the popular DH5. It is T1 phage resistant and endA deficient for high-quality plasmid preparations.
  • High efficiency strain ideal for a wide variety of applications
  • In a wide variety of sizes, including single-use vials, 200 µl vials, 96- and 384-well plates
  • No dry ice surcharge on competent cell shipments
  • Outgrowth medium included
  • Free of animal products