NcoI

Supplier: New England Biolabs (NEB)

R0193M R0193T
CA101228-982EA 447 CAD
CA101228-982 CA101228-986
NcoI
Nucleic Acid Reagents Restriction Enzymes

An E.coli strain that carries the NcoI gene from Nocardia corallina (ATCC 19070).

Order Now

SPECIFICATIONS

Environmentally Preferable
Application
Enzyme name NcoI
Source

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR