Mung bean Nuclease
Supplier: New England Biolabs (NEB)
M0250L M0250S
CA101228-274EA 458 CAD
CA101228-274 CA101228-276
Mung bean Nuclease
Enzymes
A single-strand specific DNA and RNA endonuclease which will degrade single-stranded extensions from the ends of DNA and RNA molecules, leaving blunt, ligatable ends.