Low Molecular Weight DNA Ladder, New England Biolabs
Supplier: New England Biolabs (NEB)
N3233L
N3233S
CA101228-496EA
449
CAD
CA101228-496
CA101228-498
Low Molecular Weight DNA Ladder, New England Biolabs
Electrophoresis Reagents
Molecular Weight Markers
A proprietary plasmid is digested to completion with appropriate restriction enzymes to yield 11 bands suitable for use as molecular weight standards for both agarose and polyacrylamide gel electrophoresis
This digested DNA includes fragments ranging from 25-766 base pairs. The 200 base pair band has increased intensity to serve as a reference point. Comes supplied with 1 vial of Gel Loading Dye, Purple (6X), no SDS.