LONGAMP® Taq 2X Master Mix, New England Biolabs
Supplier: New England Biolabs (NEB)
M0287L M0287S
CA101444-752EA 942 CAD
CA101444-752 CA101444-754
LONGAMP® Taq 2X Master Mix, New England Biolabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
The LongAmp® Taq 2X Master Mix combines high quality NEB recombinant Taq DNA Polymerase with a trace amount of Deep VentR™ DNA Polymerase
The 3´→5´ exonuclease activity of Deep Vent DNA Polymerase increases the fidelity and robust amplification of Taq DNA Polymerase. This ready-to-use mix offers two fold higher fidelity than Taq DNA Polymerase alone. A wide range of PCR products can be generated, up to 30 kb from lambda or human genomic DNA.