Lambda Protein Phosphatase
Supplier: New England Biolabs (NEB)
P0753L P0753S
CA101227-764EA 1085 CAD
CA101227-764 CA101227-766
Lambda Protein Phosphatase
Enzymes
Lambda Protein Phosphatase (Lambda PP) is a Mn2+-dependent protein phosphatase with activity towards phosphorylated serine, threonine and tyrosine residues
Lambda-PPase is active on phosphorylated histidine residues. It is the 221 amino-acid product of the ORF221 open reading frame on bacteriophage lambda.