Lambda DNA-Hind III Digest, New England Biolabs
Supplier: New England Biolabs (NEB)
N3012L
N3012S
CA101228-434EA
466
CAD
CA101228-434
CA101228-436
Lambda DNA-Hind III Digest, New England Biolabs
Electrophoresis Reagents
Molecular Weight Markers
The HindIII digest of lambda DNA (cI857ind1 Sam 7) yields 8 fragments suitable for use as molecular weight standards for agarose gel electrophoresis
The approximate mass of DNA in each of the bands is provided (assuming a 1.0 μg load) for approximating the mass of DNA in comparably intense samples of similar size. Comes supplied with 1 vial of Gel Loading Dye, Purple (6X), no SDS.