Hot Start Taq 2X Master Mix, New England Biolabs
Supplier: New England Biolabs (NEB)
M0496L M0496S
CA102715-952EA 396 CAD
CA102715-952 CA102715-954
Hot Start Taq 2X Master Mix, New England Biolabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Hot Start Taq 2X Master Mix is an optimized ready-to-use solution containing Hot Start Taq DNA Polymerase, dNTPs, MgCl2, KCl and stabilizers
It is ideally suited to routine PCR applications from templates including pure DNA solutions, bacterial colonies, and cDNA products. It is recommended for amplification up to 5 kb.