Hemo KlenTaq, New England Biolabs
Supplier: New England Biolabs (NEB)
M0332L
M0332S
CA101449-330EA
758
CAD
CA101449-330
CA101449-332
Hemo KlenTaq, New England Biolabs
Nucleic Acid Reagents
End-point PCR Enzymes and Kits
Hemo KlenTaq is a truncated version of Taq DNA Polymerase, lacking the first 280 amino acids
Hemo KlenTaq also contains mutations that make it resistant to inhibitors present in whole blood. It can amplify DNA from whole blood samples from humans and mice, without the need for DNA extraction. Hemo KlenTaq tolerates up to 20% whole blood in a 25 µl reaction (30% in a 50 µl reaction). Hemo KlenTaq works well with most common anticoagulants, including heparin, citrate and EDTA.