GpC Methyltransferase (M.CviPI), New England Biolabs
Supplier: New England Biolabs (NEB)
M0227L M0227S M0227L M0227S
101444-738EA 573.54 CAD
101444-738 101444-740 CA101444-738 CA101444-740
GpC Methyltransferase (M.CviPI), New England Biolabs
Enzymes
The GpC Methyltransferase (M.CviPI) methylates all cytosine residues (C5) within the double-stranded dinucleotide recognition sequence 5´...GC...3´.