G(5')ppp(5') RNA Cap Structure Analog, New England Biolabs
Supplier: New England Biolabs (NEB)
S1407L S1407S
CA101228-114EA 853 CAD
CA101228-114 CA101228-116
G(5')ppp(5') RNA Cap Structure Analog, New England Biolabs
Nucleic Acid Reagents Nucleic Acid Modification and Labelling
The 5´ terminal m7G cap present on most eukaryotic mRNAs promotes translation in vitro at the initiation level.
For most RNAs, elimination of the cap structure causes a loss of stability, especially against exonuclease degradation, and a decrease in the formation of the initiation complex of mRNAs for protein synthesis. Certain prokaryotic mRNAs containing a 5´ terminal cap structure are translated as efficiently as or more efficiently than eukaryotic mRNAs in a eukaryotic cell-free protein synthesizing system. Also a cap requirement has been observed for splicing eukaryotic substrate RNAs. A method using E. coli RNA Polymerase primed with m7G(5´)ppp(5´)G or m7G(5´)ppp(5´)A for an efficient in vitro synthesis of capped RNAs has been developed by Contreas. Larger amounts of capped RNAs are produced by transcription systems using SP6 RNA polymerase primed with m7G(5´)ppp(5´)G.
Order Now
SPECIFICATIONS
Environmentally Preferable
Application
Learn more