Exonuclease T
Supplier: New England Biolabs (NEB)
M0265L
M0265S
CA101228-314EA
505
CAD
CA101228-314
CA101228-316
Exonuclease T
Enzymes
Exonuclease T (Exo T), also known as RNase T, is a single-stranded RNA or DNA specific nuclease that requires a free 3´ terminus and removes nucleotides in the 3´→ 5´ direction
Exonuclease T can be used to generate blunt ends from RNA or DNA molecules that have 3´ extensions.