E. coli Poly (A) Polymerase, New England Biolabs
Supplier: New England Biolabs (NEB)
M0276L M0276S
CA101417-970EA 489 CAD
CA101417-970 CA101417-972
E. coli Poly (A) Polymerase, New England Biolabs
Nucleic Acid Reagents Nucleic Acid Modification and Labelling
E.coli Poly(A) Polymerase catalyzes the template independent addition of AMP from ATP to the 3´ end of RNA.
Order Now
SPECIFICATIONS
Environmentally Preferable
Application
Learn more