Desoxyribonuclease I (DNase I), New England Biolabs

Supplier: New England Biolabs (NEB)

M0303L M0303S
CA101228-384EA 472 CAD
CA101228-384 CA101228-386
Desoxyribonuclease I (DNase I), New England Biolabs
Enzymes

DNase I, (RNase-free) is an endonuclease that nonspecifically cleaves DNA to release di-, tri- and oligonucleotide products with 5´-phosphorylated and 3´-hydroxylated ends


DNase I acts on single- and double-stranded DNA, chromatin and RNA:DNA hybrids.

Order Now

SPECIFICATIONS

Enzyme name Desoxyribonuclease I (DNase I)
Environmentally Preferable
Source
Species
CAS No. 9003-98-9
Storage conditions Freezer

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR