DNA Polymerase I Klenow Fragment, New England Biolabs
Supplier: New England Biolabs (NEB)
M0210L
M0210M
M0210S
CA101228-208EA
428
CAD
CA101228-208
CA101228-210
CA101228-212
DNA Polymerase I Klenow Fragment, New England Biolabs
Nucleic Acid Reagents
Nucleic Acid Modification and Labelling
DNA Polymerase I, Large (Klenow) Fragment was originally derived as a proteolytic product of E.coli DNA Polymerase I that retains polymerase and 3´→ 5´ exonuclease activity, but lacks 5´→ 3´ exonuclease activity.
Klenow retains the polymerization fidelity of the holoenzyme without degrading 5´ termini.