DNA Polymerase I (E. coli), New England Biolabs
Supplier: New England Biolabs (NEB)
M0209L M0209S
CA101228-204EA 476 CAD
CA101228-204 CA101228-206
DNA Polymerase I (E. coli), New England Biolabs
Nucleic Acid Reagents Nucleic Acid Modification and Labelling
DNA Polymerase I (E coli) is a DNA-dependent DNA polymerase with inherent 3´→ 5´ and 5´→ 3´ exonuclease activities
The 5´→ 3´ exonuclease activity removes nucleotides ahead of the growing DNA chain, allowing nick-translation.